Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSN SIRNA Assays ›
Gene Symbol:
GSN
Gene Name:
gelsolin
Gene Aliases:
ADF, AGEL
Chromosome Location:
Chr.9: 121201082 - 121332844 on Build GRCh38
UniGene ID:
Hs.522373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
242585
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000177.4
NP_000168.1
2
375
NM_001127662.1
NP_001121134.1
2
288
NM_001127663.1
NP_001121135.2
4
493
NM_001127664.1
NP_001121136.1
4
460
NM_001127665.1
NP_001121137.1
4
455
NM_001127666.1
NP_001121138.1
4
452
NM_001127667.1
NP_001121139.1
4
443
NM_001258029.1
NP_001244958.1
2
275
NM_001258030.1
NP_001244959.1
2
352
NM_198252.2
NP_937895.1
3
381
XM_005251943.1
XP_005252000.1
3
479
XM_005251944.1
XP_005252001.1
3
470
XM_005251945.3
XP_005252002.1
3
383
XM_006717079.1
XP_006717142.1
3
317
XM_011518584.1
XP_011516886.1
4
470
XM_011518585.1
XP_011516887.1
2
290
XM_011518586.1
XP_011516888.1
5
493
XM_011518587.2
XP_011516889.1
5
535
XM_011518588.2
XP_011516890.1
5
497
XM_011518589.2
XP_011516891.1
5
493
XM_011518590.2
XP_011516892.1
5
445
XM_011518591.2
XP_011516893.1
10
1537
XM_011518592.1
XP_011516894.1
4
359
XM_011518593.1
XP_011516895.1
5
505
XM_017014643.1
XP_016870132.1
11
1688
XM_017014644.1
XP_016870133.1
11
1622
XM_017014645.1
XP_016870134.1
3
434
XM_017014646.1
XP_016870135.1
6
605
XM_017014647.1
XP_016870136.1
12
1821
XM_017014648.1
XP_016870137.1
5
432
GenBank mRNA
AK096280.1
-
3
354
AK125819.1
-
11
1184
AK295201.1
-
3
297
AK295572.1
-
4
414
AK297870.1
-
2
233
AK299453.1
-
2
340
AK300631.1
-
4
427
AK304629.1
-
296
AK304675.1
-
1
329
AK307859.1
-
2
239
AK315494.1
-
2
346
BC026033.1
-
2
319
4661084
BU151663.1
-
3
281
BX647999.1
-
2
277
DA294394.1
-
3
381
DA394782.1
-
4
448
DA450851.1
-
2
275
DA823796.1
-
2
275
DB170452.1
-
4
398
DB250757.1
-
2
375
DC316173.1
-
4
404
DC341948.1
-
4
418
DC370937.1
-
4
418
DC378873.1
-
2
353
X04412.1
-
2
328
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis, engulfment
aging
positive regulation of gene expression
oligodendrocyte development
striated muscle atrophy
extracellular matrix disassembly
actin filament polymerization
protein destabilization
tissue regeneration
sequestering of actin monomers
cellular protein metabolic process
actin nucleation
response to ethanol
negative regulation of viral entry into host cell
phosphatidylinositol-mediated signaling
actin filament severing
barbed-end actin filament capping
positive regulation of actin nucleation
response to folic acid
actin filament capping
cilium morphogenesis
cellular response to cadmium ion
regulation of podosome assembly
actin filament reorganization
renal protein absorption
hepatocyte apoptotic process
positive regulation of keratinocyte apoptotic process
regulation of wound healing, spreading of epidermal cells
regulation of establishment of T cell polarity
regulation of plasma membrane raft polarization
regulation of receptor clustering
positive regulation of protein processing in phagocytic vesicle
amyloid fibril formation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
actin binding
calcium ion binding
protein binding
protein domain specific binding
myosin II binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline