Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178449.3 | NP_848544.1 | 1 | 199 | ||
GenBank mRNA | AY037555.1 | - | 1 | 199 | ||
AY048588.1 | - | 1 | 100 | |||
BC093995.1 | - | 1 | 138 | 7939840 | ||
BC093997.1 | - | 1 | 142 | 7939842 |