Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016441.2 | NP_057525.1 | 2 | 545 | ||
XM_011532898.2 | XP_011531200.1 | 3 | 666 | |||
XM_011532899.2 | XP_011531201.1 | 3 | 667 | |||
XM_011532901.2 | XP_011531203.1 | 3 | 667 | |||
XM_017004258.1 | XP_016859747.1 | 3 | 666 | |||
XM_017004259.1 | XP_016859748.1 | 2 | 684 | |||
XM_017004260.1 | XP_016859749.1 | 2 | 541 | |||
XM_017004261.1 | XP_016859750.1 | 3 | 323 | |||
XM_017004262.1 | XP_016859751.1 | 2 | 206 | |||
GenBank mRNA | AB209139.1 | - | 2 | 545 | ||
AF167706.1 | - | 2 | 518 | |||
AY358371.1 | - | 2 | 846 | |||
BC111989.1 | - | 2 | 516 | 8327457 | ||
BC113371.1 | - | 2 | 516 | 8322423 |