Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016468.6 | NP_057552.1 | 2 | 237 | ||
GenBank mRNA | AF151037.1 | - | 2 | 184 | ||
AF226729.1 | - | 2 | 211 | |||
AK290743.1 | - | 2 | 185 | |||
BC001702.2 | - | 2 | 137 | 2966935 | ||
BM799209.1 | - | 2 | 206 | |||
CR457199.1 | - | 2 | 93 | |||
FJ460510.1 | - | 2 | 237 | |||
FJ460511.1 | - | 2 | 237 |
Gene Symbol: SYNJ2BP-COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202548.2 | NP_001189477.1 | 4 | 448 | ||
GenBank mRNA | FY211163.1 | - | 3 | 205 | ||
FY213836.1 | - | 4 | 342 |