Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAS3 SIRNA Assays ›
Gene Symbol:
BCAS3
Gene Name:
BCAS3, microtubule associated cell migration factor
Gene Aliases:
GAOB1, MAAB
Chromosome Location:
Chr.17: 60677811 - 61392838 on Build GRCh38
UniGene ID:
Hs.655028
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
243748
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099432.1
NP_001092902.1
25
2792
NM_001320470.1
NP_001307399.1
26
2904
NM_017679.3
NP_060149.3
24
2747
XM_005257472.1
XP_005257529.1
26
2809
XM_011524939.1
XP_011523241.1
29
3178
XM_011524940.1
XP_011523242.1
28
3133
XM_011524941.1
XP_011523243.1
27
3043
XM_011524942.2
XP_011523244.1
27
2899
XM_011524943.2
XP_011523245.1
26
2854
XM_011524944.1
XP_011523246.1
26
2833
XM_011524945.2
XP_011523247.1
25
2788
XM_017024783.1
XP_016880272.1
30
3269
XM_017024784.1
XP_016880273.1
28
3112
XM_017024785.1
XP_016880274.1
29
3203
XM_017024786.1
XP_016880275.1
28
3088
XM_017024787.1
XP_016880276.1
28
3040
XM_017024788.1
XP_016880277.1
27
2974
XM_017024789.1
XP_016880278.1
27
2924
XM_017024790.1
XP_016880279.1
27
2900
XM_017024791.1
XP_016880280.1
25
2764
XM_017024792.1
XP_016880281.1
26
2834
XM_017024793.1
XP_016880282.1
25
2789
XM_017024796.1
XP_016880285.1
17
2169
GenBank mRNA
AF260268.1
-
9
1113
AF361219.1
-
25
2686
AJ511332.1
-
3
198
AK000135.1
-
11
1291
AK022526.1
-
16
1996
AK023054.1
-
16
2094
AK025510.1
-
25
2708
AK225757.1
-
26
2865
AK314744.1
-
24
2692
AL831895.1
-
12
1329
BC001250.1
-
10
1234
BC013841.1
-
4
1181
BC029983.1
-
4
1181
BC036280.1
-
4
1284
BC117275.1
-
25
2714
40125826
BC143386.1
-
25
2714
9051892
BC143387.1
-
24
2669
9051893
BX648103.1
-
1
3743
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription, DNA-templated
Golgi organization
positive regulation of endothelial cell migration
vesicle-mediated transport
microtubule organizing center organization
negative regulation of GTPase activity
tube formation
response to starvation
positive regulation of catalytic activity
positive regulation of GTPase activity
positive regulation of transcription from RNA polymerase II promoter
positive regulation of filopodium assembly
negative regulation of focal adhesion assembly
cellular response to estrogen stimulus
positive regulation of intracellular protein transport
activation of GTPase activity
regulation of establishment of cell polarity
positive regulation of actin cytoskeleton reorganization
chromatin binding
protein binding
transcription factor binding
acetyltransferase activator activity
histone acetyltransferase binding
nuclear hormone receptor binding
histone binding
beta-tubulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-xm8cg:80/10.239.167.213:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0