Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137604.2 | NP_001131076.1 | 13 | 2895 | ||
NM_001282772.1 | NP_001269701.1 | 15 | 2713 | |||
NM_001282774.1 | NP_001269703.1 | 12 | 2514 | |||
NM_001282776.1 | NP_001269705.1 | 11 | 2615 | |||
NM_001282777.1 | NP_001269706.1 | 15 | 3155 | |||
NM_001282779.1 | NP_001269708.1 | 14 | 2992 | |||
NM_019014.5 | NP_061887.2 | 14 | 3063 | |||
GenBank mRNA | AK001678.1 | - | 14 | 2535 | ||
AK022533.1 | - | 13 | 2345 | |||
AK022890.1 | - | 7 | 1226 | |||
AK025574.1 | - | 3 | 571 | |||
AK128044.1 | - | 14 | 3063 | |||
AK294027.1 | - | 14 | 2464 | |||
AK301178.1 | - | 11 | 2087 | |||
AK302814.1 | - | 15 | 2713 | |||
BC004882.2 | - | 5 | 825 | |||
BC073857.1 | - | 5 | 759 | |||
BC096748.1 | - | 5 | 860 | |||
BC110833.1 | - | 12 | 2203 | 6053210 | ||
BC150139.1 | - | 6 | 1269 | |||
BX647360.1 | - | 15 | 2881 | |||
BX647683.1 | - | 14 | 2442 |