Search Thermo Fisher Scientific
Gene Symbol: CPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016352.3 | NP_057436.2 | 3, 4 | 340 | ||
GenBank mRNA | AF095719.1 | - | 3, 4 | 300 | ||
AK290993.1 | - | 3, 4 | 340 | |||
AK298553.1 | - | 3, 4 | 313 | |||
AY358699.1 | - | 3, 4 | 301 | |||
BC052289.1 | - | 3, 4 | 321 | 6106874 | ||
DC368360.1 | - | 3, 4 | 340 |