Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170938.1 | NP_001164409.1 | 7 | 1120 | ||
NM_001253389.1 | NP_001240318.1 | 5 | 986 | |||
NM_016368.4 | NP_057452.1 | 8 | 1282 | |||
NR_045573.1 | 7 | 1088 | ||||
NR_045574.1 | 5 | 952 | ||||
XM_011528059.1 | XP_011526361.1 | 7 | 1361 | |||
GenBank mRNA | AF086921.1 | - | 8 | 1101 | ||
AF207640.1 | - | 8 | 1094 | |||
AF220530.1 | - | 8 | 1114 | |||
AK001325.1 | - | 7 | 1068 | |||
AK021526.1 | - | 8 | 1114 | |||
AK092179.1 | - | 7 | 986 | |||
AK092433.1 | - | 4 | 1201 | |||
AK093461.1 | - | 5 | 901 | |||
AK296022.1 | - | 7 | 950 | |||
AL137749.1 | - | 5 | 986 | |||
BC018952.2 | - | 8 | 1097 | 4301272 | ||
BC066902.1 | - | 8 | 1112 | 5262041 |