Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 14 | 4542 | ||
XM_011512928.2 | XP_011511230.1 | 14 | 4633 | |||
GenBank mRNA | AB033094.1 | - | 9 | 2512 | ||
AK001770.1 | - | 10 | 1570 | |||
AK022542.1 | - | 2 | 347 | |||
AK026003.1 | - | 9 | 2371 | |||
AK304269.1 | - | 14 | 4510 | |||
AY134858.1 | - | 9 | 3715 | |||
BC032804.1 | - | 1 | 258 | |||
BX648758.1 | - | 10 | 3860 | |||
DQ063584.1 | - | 11 | 4006 | |||
DQ063585.1 | - | 11 | 4006 |