Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182491.2 | NP_872297.2 | 4 | 475 | ||
XM_017012774.1 | XP_016868263.1 | 4 | 475 | |||
GenBank mRNA | AK027118.1 | - | 4 | 417 | ||
AK290170.1 | - | 4 | 434 | |||
BC016963.1 | - | 4 | 410 | |||
BC029558.1 | - | 4 | 418 | 5263788 |