Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM9 SIRNA Assays ›
Gene Symbol:
TMEM9
Gene Name:
transmembrane protein 9
Gene Aliases:
DERM4, TMEM9A
Chromosome Location:
Chr.1: 201134771 - 201171625 on Build GRCh38
UniGene ID:
Hs.181444
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24460
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288564.1
NP_001275493.1
5, 6
524
NM_001288565.1
NP_001275494.1
4, 5
1027
NM_001288566.1
NP_001275495.1
5, 6
625
NM_001288567.1
NP_001275496.1
5, 6
839
NM_001288568.1
NP_001275497.1
5, 6
677
NM_001288569.1
NP_001275498.1
5, 6
538
NM_001288570.1
NP_001275499.1
5, 6
551
NM_001288571.1
NP_001275500.1
5, 6
617
NM_016456.4
NP_057540.1
5, 6
542
NR_109920.1
6, 7
769
NR_109921.1
4, 5
996
XM_011509385.1
XP_011507687.1
5, 6
601
XM_011509386.2
XP_011507688.1
5, 6
576
XM_011509387.2
XP_011507689.1
5, 6
464
GenBank mRNA
AB013909.1
-
4, 5
395
AF151020.1
-
6, 7
514
AK054883.1
-
5, 6
497
AK075335.1
-
5, 6
452
AK091726.1
-
4, 5
912
AK094588.1
-
1, 2
1845
AK303843.1
-
5, 6
563
AL136600.1
-
5, 6
481
AL516746.3
-
5, 6
716
AY138587.1
-
5, 6
453
AY359012.1
-
5, 6
516
BC001106.2
-
5, 6
466
3506442
BE782946.1
-
5, 6
503
BX337142.2
-
4, 5
880
CR457162.1
-
4, 5
395
CR533467.1
-
4, 5
401
DA141558.1
-
5, 6
524
DA364005.1
-
5, 6
542
DB031447.1
-
5, 6
518
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transport
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-9xb8z:80/10.239.165.207:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0