Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPHLN1 SIRNA Assays ›
Gene Symbol:
PPHLN1
Gene Name:
periphilin 1
Gene Aliases:
CR, HSPC206, HSPC232
Chromosome Location:
Chr.12: 42326126 - 42448621 on Build GRCh38
UniGene ID:
Hs.444157
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24481
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPHLN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143787.1
NP_001137259.1
3, 4
232
NM_001143788.1
NP_001137260.1
4, 5
397
NM_001143789.1
NP_001137261.1
3, 4
232
NM_016488.6
NP_057572.5
4, 5
397
NM_201438.1
NP_958846.1
4, 5
275
NM_201439.1
NP_958847.1
4, 5
397
NM_201440.1
NP_958848.1
4, 5
275
NM_201515.1
NP_958923.1
5, 6
440
XM_005268962.3
XP_005269019.1
3, 4
251
XM_011538459.2
XP_011536761.1
5, 6
418
XM_011538465.2
XP_011536767.1
4, 5
375
XM_011538467.2
XP_011536769.1
4, 5
257
XM_011538470.2
XP_011536772.1
4, 5
275
XM_011538471.2
XP_011536773.1
3, 4
251
XM_017019434.1
XP_016874923.1
5, 6
418
XM_017019435.1
XP_016874924.1
5, 6
417
XM_017019436.1
XP_016874925.1
5, 6
418
XM_017019437.1
XP_016874926.1
4, 5
375
XM_017019438.1
XP_016874927.1
4, 5
375
XM_017019439.1
XP_016874928.1
5, 6
418
XM_017019440.1
XP_016874929.1
5, 6
418
XM_017019441.1
XP_016874930.1
4, 5
375
XM_017019442.1
XP_016874931.1
4, 5
375
XM_017019443.1
XP_016874932.1
5, 6
418
XM_017019444.1
XP_016874933.1
4, 5
257
XM_017019445.1
XP_016874934.1
3, 4
251
XM_017019446.1
XP_016874935.1
3, 4
250
XM_017019447.1
XP_016874936.1
4, 5
275
XM_017019448.1
XP_016874937.1
4, 5
275
XM_017019449.1
XP_016874938.1
3, 4
251
XM_017019450.1
XP_016874939.1
3, 4
251
XM_017019451.1
XP_016874940.1
4, 5
275
XM_017019452.1
XP_016874941.1
5, 6
418
XM_017019453.1
XP_016874942.1
4, 5
275
GenBank mRNA
AF151066.1
-
4, 5
250
AK055690.1
-
4, 5
385
AK129910.1
-
5, 6
440
AK299951.1
-
3, 4
214
AK303612.1
-
3, 4
232
AK308022.1
-
4, 5
371
AY039238.1
-
4, 5
379
AY157850.1
-
5, 6
436
BC025306.1
-
3, 4
199
BC039832.1
-
4, 5
242
6051282
BC040449.1
-
4, 5
242
CF242917.1
-
3, 4
247
DC424443.1
-
3, 4
217
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
keratinization
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0