Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136053.2 | NP_001129525.1 | 11 | 1281 | ||
NM_001142646.2 | NP_001136118.1 | 9 | 1117 | |||
NR_073377.1 | 10 | 1220 | ||||
XM_006713495.3 | XP_006713558.1 | 12 | 1462 | |||
XM_006713496.2 | XP_006713559.1 | 11 | 1139 | |||
XM_006713497.3 | XP_006713560.1 | 12 | 1506 | |||
XM_011512422.1 | XP_011510724.1 | 11 | 1055 | |||
XM_017005707.1 | XP_016861196.1 | 12 | 1245 | |||
XM_017005708.1 | XP_016861197.1 | 12 | 1275 | |||
XM_017005709.1 | XP_016861198.1 | 11 | 1315 | |||
XM_017005710.1 | XP_016861199.1 | 11 | 1154 | |||
GenBank mRNA | AB037108.1 | - | 10 | 1205 | ||
AF289568.1 | - | 9 | 1064 | |||
AK056759.1 | - | 1 | 1291 | |||
BC017058.1 | - | 10 | 989 | 3904719 | ||
BC050711.2 | - | 11 | 1175 | 6168632 |