Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 2 | 310 | ||
NR_033249.1 | 2 | 310 | ||||
XM_017023136.1 | XP_016878625.1 | 2 | 309 | |||
XM_017023137.1 | XP_016878626.1 | 2 | 252 | |||
GenBank mRNA | AK311978.1 | - | 2 | 219 | ||
AV696698.1 | - | 2 | 207 | |||
AY533031.1 | - | 1 | 69 | |||
BC000790.1 | - | 1 | 234 | 3451361 | ||
BC009065.1 | - | 1 | 269 | |||
BC020922.1 | - | 1 | 273 | 4247157 | ||
BF798008.1 | - | 2 | 181 | |||
BG708388.1 | - | 2 | 343 | |||
BT009827.1 | - | 2 | 213 | |||
CN283805.1 | - | 2 | 200 | |||
CN283809.1 | - | 2 | 343 | |||
CR457072.1 | - | 2 | 213 | |||
D00723.1 | - | 1 | 305 | |||
M69175.1 | - | 2 | 237 |