Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000777.4 | NP_000768.1 | 13 | 1658 | ||
NM_001291829.1 | NP_001278758.1 | 14 | 2158 | |||
NM_001291830.1 | NP_001278759.1 | 14 | 1918 | |||
NR_033807.2 | 13 | 3262 | ||||
XM_011515843.1 | XP_011514145.1 | 14 | 1775 | |||
XM_017011781.1 | XP_016867270.1 | 10 | 1300 | |||
XM_017011782.1 | XP_016867271.1 | 8 | 1097 | |||
GenBank mRNA | AA076026.1 | - | 2 | 245 | ||
BX537676.1 | - | 13 | 3233 |