Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYP4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000778.3 | NP_000769.2 | 12 | 2091 | ||
NM_001319155.1 | NP_001306084.1 | 12 | 1995 | |||
NR_134988.1 | 10 | 1745 | ||||
NR_134989.1 | 11 | 1936 | ||||
NR_134990.1 | 11 | 1930 | ||||
NR_134991.1 | 11 | 1917 | ||||
NR_134992.1 | 8 | 1546 | ||||
NR_134993.1 | 9 | 1680 | ||||
NR_134994.1 | 11 | 1952 | ||||
XM_017000464.1 | XP_016855953.1 | 10 | 1789 | |||
XM_017000465.1 | XP_016855954.1 | 12 | 2010 | |||
GenBank mRNA | D26481.1 | - | 12 | 2054 | ||
L04751.1 | - | 12 | 2072 | |||
S67580.1 | - | 12 | 2082 | |||
S67581.1 | - | 12 | 2081 |
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270767.3 | XP_005270824.1 | 10 | 1789 |