Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021154.4 | NP_066977.1 | 7 | 993 | ||
NM_058179.3 | NP_478059.1 | 7 | 993 | |||
GenBank mRNA | AF113132.1 | - | 7 | 890 | ||
AK025460.1 | - | 7 | 927 | |||
AK295222.1 | - | 7 | 993 | |||
AK295310.1 | - | 8 | 882 | |||
AY131232.1 | - | 7 | 902 | |||
BC000971.1 | - | 7 | 915 | 3447262 | ||
BC004863.2 | - | 7 | 924 | 3938160 | ||
BC016645.2 | - | 7 | 903 | 4342332 | ||
BC018129.1 | - | 7 | 924 | 3883843 | ||
BT006840.1 | - | 7 | 857 |