Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016579.3 | NP_057663.1 | 2, 3 | 343 | ||
GenBank mRNA | AF161254.1 | - | 2, 3 | 342 | ||
AK222623.1 | - | 2, 3 | 327 | |||
AK315656.1 | - | 2, 3 | 328 | |||
AL136652.1 | - | 2, 3 | 332 | |||
AL365455.1 | - | 2, 3 | 315 | |||
AY358420.1 | - | 2, 3 | 309 | |||
BC000668.2 | - | 2, 3 | 314 | 3347569 | ||
BC007083.1 | - | 2, 3 | 297 | 4076237 | ||
CB141050.1 | - | 2, 3 | 343 | |||
CR457174.1 | - | 2, 3 | 277 |