Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033257.3 | NP_150282.2 | 2 | 296 | ||
GenBank mRNA | AF228708.1 | - | 2 | 276 | ||
AK001528.1 | - | 2 | 237 | |||
AK307210.1 | - | 2 | 232 | |||
BC000682.2 | - | 2 | 232 | 3349309 | ||
CR456361.1 | - | 2 | 201 | |||
KJ535014.1 | - | 2 | 179 |
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 2 | 331 | ||
XM_011530394.1 | XP_011528696.1 | 2 | 199 | |||
GenBank mRNA | AF228707.1 | - | 2 | 275 | ||
AK098780.1 | - | 2 | 199 | |||
AK315115.1 | - | 2 | 236 | |||
BC047039.1 | - | 2 | 224 | 5229172 | ||
BQ639188.1 | - | 2 | 331 | |||
CR456434.1 | - | 2 | 201 | |||
KJ534820.1 | - | 2 | 179 | |||
KJ534971.1 | - | 2 | 179 | |||
KJ535003.1 | - | 2 | 179 | |||
KJ535013.1 | - | 2 | 179 | |||
KJ535027.1 | - | 2 | 165 | |||
KJ535047.1 | - | 1 | 258 | |||
X96484.1 | - | 2 | 192 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 2 | 273 |