Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205254.1 | NP_001192183.1 | 7 | 1505 | ||
NM_001205255.1 | NP_001192184.1 | 5 | 688 | |||
NM_002538.3 | NP_002529.1 | 7 | 1735 | |||
XM_017008913.1 | XP_016864402.1 | 6 | 1343 | |||
XM_017008914.1 | XP_016864403.1 | 6 | 1318 | |||
GenBank mRNA | AB451306.1 | - | 6 | 1299 | ||
AB451437.1 | - | 6 | 1299 | |||
AK290697.1 | - | 7 | 1467 | |||
AK311037.1 | - | 3 | 360 | |||
AK313501.1 | - | 7 | 1477 | |||
BC029886.1 | - | 7 | 1477 | 5179203 | ||
FJ786083.1 | - | 5 | 664 | |||
FJ786084.1 | - | 4 | 502 | |||
GQ225098.1 | - | 6 | 912 | |||
GQ402517.1 | - | 6 | 1343 | |||
U49184.1 | - | 7 | 1466 | |||
U53823.1 | - | 7 | 1736 |
Gene Symbol: LOC647859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026578.1 | 3 | 286 | |||
GenBank mRNA | DC407815.1 | - | 3 | 286 |