Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205254.1 | NP_001192183.1 | 7 | 1559 | ||
NM_001205255.1 | NP_001192184.1 | 5 | 742 | |||
NM_002538.3 | NP_002529.1 | 7 | 1789 | |||
XM_017008913.1 | XP_016864402.1 | 6 | 1397 | |||
XM_017008914.1 | XP_016864403.1 | 6 | 1372 | |||
GenBank mRNA | AB451306.1 | - | 6 | 1353 | ||
AB451437.1 | - | 6 | 1353 | |||
AK290697.1 | - | 7 | 1521 | |||
AK311037.1 | - | 3 | 414 | |||
AK313501.1 | - | 7 | 1531 | |||
BC029886.1 | - | 7 | 1531 | 5179203 | ||
FJ786083.1 | - | 5 | 718 | |||
FJ786084.1 | - | 4 | 556 | |||
GQ225098.1 | - | 6 | 966 | |||
GQ402517.1 | - | 6 | 1397 | |||
U49184.1 | - | 7 | 1520 | |||
U53823.1 | - | 7 | 1790 |
Gene Symbol: LOC647859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026578.1 | 3 | 340 | |||
GenBank mRNA | DC407815.1 | - | 3 | 340 |