Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101802.1 | NP_001095272.1 | 4, 5 | 710 | ||
NM_016621.3 | NP_057705.3 | 4, 5 | 710 | |||
XM_005252965.4 | XP_005253022.1 | 5, 6 | 399 | |||
XM_011520173.2 | XP_011518475.1 | 5, 6 | 519 | |||
XM_011520174.2 | XP_011518476.1 | 6, 7 | 493 | |||
XM_011520175.2 | XP_011518477.1 | 6, 7 | 493 | |||
XM_011520179.2 | XP_011518481.1 | 5, 6 | 513 | |||
XM_017017891.1 | XP_016873380.1 | 5, 6 | 519 | |||
XM_017017892.1 | XP_016873381.1 | 5, 6 | 519 | |||
XM_017017897.1 | XP_016873386.1 | 5, 6 | 591 | |||
XM_017017898.1 | XP_016873387.1 | 5, 6 | 449 | |||
XM_017017899.1 | XP_016873388.1 | 6, 7 | 583 | |||
XM_017017900.1 | XP_016873389.1 | 5, 6 | 538 | |||
XM_017017902.1 | XP_016873391.1 | 5, 6 | 519 | |||
XM_017017903.1 | XP_016873392.1 | 2, 3 | 169 | |||
XM_017017906.1 | XP_016873395.1 | 2, 3 | 169 | |||
XM_017017907.1 | XP_016873396.1 | 5, 6 | 519 | |||
XM_017017908.1 | XP_016873397.1 | 6, 7 | 636 | |||
GenBank mRNA | AB051483.1 | - | 4, 5 | 454 | ||
AK021530.1 | - | 4, 5 | 358 | |||
AK023258.1 | - | 6, 7 | 471 | |||
AY090777.1 | - | 2, 3 | 86 | |||
BC015714.2 | - | 5, 6 | 374 | 3908900 |