Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006855.3 | NP_006846.1 | 4 | 609 | ||
NM_016657.2 | NP_057839.1 | 4 | 609 | |||
GenBank mRNA | AK129888.1 | - | 4 | 575 | ||
AK222930.1 | - | 4 | 589 | |||
AK223238.1 | - | 4 | 566 | |||
AK292102.1 | - | 4 | 601 | |||
AL035081.1 | - | 4 | 562 | |||
AL035082.1 | - | 4 | 593 | |||
BC001277.1 | - | 4 | 586 | 3462392 | ||
BC096722.1 | - | 4 | 543 | 40003712 | ||
BC098131.1 | - | 4 | 543 | 40003710 | ||
BC098156.1 | - | 4 | 543 | 40003711 | ||
BC098336.1 | - | 4 | 543 | 40003709 | ||
CR456509.1 | - | 4 | 484 |