Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 1 | 22 | |||
NR_029426.1 | 4 | 314 | ||||
GenBank mRNA | AK094021.1 | - | 5 | 720 | ||
AK097091.1 | - | 5 | 694 | |||
AK289832.1 | - | 4 | 396 | |||
AK315849.1 | - | 4 | 387 | |||
X75940.1 | - | 1 | 22 | |||
X83300.1 | - | 2 | 137 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 4 | 277 | |||
GenBank mRNA | AK290693.1 | - | 4 | 244 | ||
BC147000.1 | - | 4 | 231 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 4 | 418 | |||
GenBank mRNA | BC034017.1 | - | 4 | 419 | ||
BC067351.1 | - | 2 | 116 | |||
BC140728.1 | - | 3 | 241 | |||
BC171739.1 | - | 3 | 241 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027386.2 | 4 | 396 | |||
GenBank mRNA | BT006760.1 | - | 2 | 71 | ||
DT218991.1 | - | 4 | 316 | |||
X83299.1 | - | 4 | 347 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 2 | 128 | |||
GenBank mRNA | AW500063.1 | - | 2 | 61 | ||
BC104473.1 | - | 2 | 128 | |||
CF141910.1 | - | 2 | 134 |
Gene Symbol: LOC100133050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027503.1 | 2 | 48 | |||
GenBank mRNA | BI830432.1 | - | 2 | 48 | ||
BQ109458.1 | - | 2 | 48 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 5 | 390 | |||
GenBank mRNA | AK289851.1 | - | 4 | 316 | ||
CN312642.1 | - | 2 | 180 | |||
X83301.1 | - | 5 | 390 |