Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123168.2 | NP_001116640.1 | 3 | 1148 | ||
NM_001317901.1 | NP_001304830.1 | 3 | 368 | |||
NR_134239.1 | 4 | 428 | ||||
XM_006711526.3 | XP_006711589.1 | 4 | 1122 | |||
XM_011509965.2 | XP_011508267.1 | 4 | 1031 | |||
XM_011509966.2 | XP_011508268.1 | 3 | 1163 | |||
XM_011509967.1 | XP_011508269.1 | 3 | 1043 | |||
XM_017002233.1 | XP_016857722.1 | 4 | 1028 | |||
GenBank mRNA | BC035696.1 | - | 3 | 715 | 5541342 | |
BC137545.1 | - | 3 | 679 | 9021545 | ||
BC146978.1 | - | 3 | 1147 | 9056906 | ||
BC146992.1 | - | 3 | 1146 | 9056920 | ||
BI916384.1 | - | 3 | 369 |
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 3 | 1127 | ||
NM_001320149.1 | NP_001307078.1 | 3 | 600 | |||
XM_011542018.1 | XP_011540320.1 | 4 | 1535 | |||
GenBank mRNA | BC046199.1 | - | 3 | 600 | 6064384 | |
BC137546.1 | - | 3 | 680 | 9021546 |