Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278928.1 | NP_001265857.1 | 1, 2 | 134 | ||
NM_017503.4 | NP_059973.4 | 1, 2 | 134 | |||
GenBank mRNA | BC014411.1 | - | 1, 2 | 103 | 4548425 | |
BI118187.1 | - | 1, 2 | 76 | |||
BM789997.1 | - | 1, 2 | 127 | |||
BP244646.1 | - | 1, 2 | 134 | |||
CR456753.1 | - | 1, 2 | 86 | |||
Z35094.1 | - | 1, 2 | 103 |