Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017995.2 | NP_001017995.1 | 4 | 466 | ||
NM_001308175.1 | NP_001295104.1 | 4 | 466 | |||
XM_017009351.1 | XP_016864840.1 | 4 | 466 | |||
GenBank mRNA | AB430862.1 | - | 4 | 295 | ||
AK128871.1 | - | 4 | 466 | |||
BC038561.1 | - | 4 | 301 | |||
DQ109556.1 | - | 4 | 301 |