Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 8 | 1258 | ||
GenBank mRNA | AB078145.1 | - | 8 | 1126 | ||
AJ505950.1 | - | 8 | 1126 | |||
AK023782.1 | - | 5 | 592 | |||
AK127135.1 | - | 8 | 1227 | |||
AL096739.1 | - | 1 | 397 | |||
BC007224.1 | - | 1 | 58 | |||
BC072450.1 | - | 4 | 558 | |||
BC098444.1 | - | 3 | 247 | |||
BG743249.1 | - | 1 | 436 |