Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 1 | 237 | ||
NM_001278923.1 | NP_001265852.1 | 1 | 237 | |||
NM_001278924.1 | NP_001265853.1 | 1 | 237 | |||
GenBank mRNA | AK223149.1 | - | 1 | 51 | ||
AK300191.1 | - | 1 | 71 | |||
AK307182.1 | - | 1 | 51 | |||
AK313626.1 | - | 1 | 100 | |||
BC008813.2 | - | 1 | 151 | 3958020 | ||
DA698972.1 | - | 1 | 220 | |||
DC381265.1 | - | 1 | 238 | |||
X16662.1 | - | 1 | 81 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717951.3 | XP_006718014.1 | 1 | 154 |