Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017565.3 | NP_060035.2 | 1, 2 | 700 | ||
XM_011524918.2 | XP_011523220.1 | 1, 2 | 540 | |||
XM_017024781.1 | XP_016880270.1 | 1, 2 | 540 | |||
GenBank mRNA | AB545606.1 | - | 1, 2 | 700 | ||
AK056789.1 | - | 1, 2 | 540 | |||
AY358197.1 | - | 1, 2 | 700 | |||
BC136686.1 | - | 1, 2 | 589 | 9020676 | ||
BC136689.1 | - | 1, 2 | 589 | 9020679 |