Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252100.1 | NP_001239029.1 | 35 | 7373 | ||
NM_001252102.1 | NP_001239031.1 | 35 | 6757 | |||
NM_001252103.1 | NP_001239032.1 | 34 | 6718 | |||
NM_017596.3 | NP_060066.2 | 34 | 7334 | |||
GenBank mRNA | AB007918.2 | - | 34 | 7334 | ||
AL137395.1 | - | 1 | 1101 | |||
BC052964.1 | - | 1 | 786 | |||
BC078676.1 | - | 6 | 2341 |