Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TEX28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205201.1 | NP_001192130.1 | 5 | 1186 | ||
NM_001586.2 | NP_001577.1 | 5 | 1180 | |||
XM_011531116.1 | XP_011529418.1 | 5 | 1224 | |||
XM_011531117.1 | XP_011529419.1 | 6 | 1293 | |||
XM_011531118.2 | XP_011529420.1 | 6 | 1421 | |||
GenBank mRNA | BC026183.1 | - | 5 | 1191 | 5266650 | |
U93720.1 | - | 5 | 1164 |