Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031157.3 | NP_112420.1 | 8, 9 | 1031 | ||
XM_005268826.1 | XP_005268883.1 | 8, 9 | 1025 | |||
GenBank mRNA | BU541100.1 | - | 5, 6 | 601 | ||
BX425482.2 | - | 8, 9 | 955 |