Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017423.2 | NP_059119.2 | 2 | 280 | ||
XM_005263062.1 | XP_005263119.1 | 2 | 280 | |||
XM_011532025.2 | XP_011530327.1 | 2 | 1027 | |||
XM_017008292.1 | XP_016863781.1 | 5 | 1200 | |||
XM_017008293.1 | XP_016863782.1 | 5 | 2989 | |||
GenBank mRNA | AJ002744.1 | - | 2 | 229 | ||
BC046129.1 | - | 2 | 258 | 4442080 | ||
BC047468.1 | - | 2 | 255 | 5271749 | ||
DB238519.1 | - | 2 | 280 |