Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 2 | 201 | ||
NR_111943.1 | 1 | 407 | ||||
NR_111944.1 | 2 | 201 | ||||
GenBank mRNA | AK311951.1 | - | 2 | 109 | ||
BC009407.2 | - | 1 | 92 | 4334552 | ||
BC019899.1 | - | 1 | 97 | 3882924 | ||
BC020453.1 | - | 1 | 96 | |||
BC022370.1 | - | 2 | 114 | 4738331 | ||
BC049824.1 | - | 1 | 89 | 6212543 | ||
BC062715.1 | - | 2 | 199 | 4068842 | ||
BC070222.1 | - | 1 | 110 | 6738383 | ||
BC071928.1 | - | 1 | 100 | 5090053 | ||
BE905890.1 | - | 1 | 77 | |||
BI597659.1 | - | 1 | 407 | |||
BM017772.1 | - | 2 | 202 | |||
BM457623.1 | - | 1 | 94 | |||
DA310472.1 | - | 1 | 407 | |||
M13932.1 | - | 2 | 110 |