Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278928.1 | NP_001265857.1 | 6 | 819 | ||
NM_017503.4 | NP_059973.4 | 6 | 822 | |||
GenBank mRNA | BC014411.1 | - | 6 | 791 | 4548425 | |
BQ672017.1 | - | 4 | 469 | |||
Z35094.1 | - | 6 | 791 |