Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL17RD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF458067.1 | - | 7, 8 | 755 | ||
AF494208.1 | - | 7, 8 | 844 | |||
AF494211.1 | - | 8, 9 | 845 | |||
AK093074.1 | - | 8, 9 | 844 | |||
AL133097.1 | - | 3, 4 | 230 | |||
AL833913.1 | - | 7, 8 | 668 | |||
AY358774.1 | - | 7, 8 | 791 | |||
AY489047.1 | - | 8, 9 | 870 | |||
BC111702.1 | - | 7, 8 | 695 | 40038598 |