Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030643.4 | NP_085146.2 | 2 | 256 | ||
NM_145660.2 | NP_663693.1 | 2 | 256 | |||
GenBank mRNA | AK313637.1 | - | 2 | 193 | ||
AY014914.1 | - | 2 | 257 | |||
AY014915.1 | - | 2 | 256 | |||
BC006276.2 | - | 2 | 166 |