Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178005.1 | NP_001171476.1 | 3 | 266 | ||
NM_017614.4 | NP_060084.2 | 3 | 266 | |||
GenBank mRNA | AF257473.1 | - | 3 | 241 | ||
AK000008.1 | - | 2 | 178 | |||
AK298298.1 | - | 3 | 223 | |||
AK315520.1 | - | 3 | 224 | |||
BC020665.1 | - | 3 | 221 | 4722608 | ||
CB140104.1 | - | 3 | 266 |