Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 17 | 2037 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 17 | 2025 | |||
GenBank mRNA | AK292534.1 | - | 17 | 1908 | ||
BC110417.1 | - | 17 | 1996 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 17 | 2025 | |||
GenBank mRNA | BC063309.1 | - | 18 | 1999 | ||
DQ309037.1 | - | 17 | 1999 |
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 17 | 2025 | |||
GenBank mRNA | AK057241.1 | - | 15 | 1503 | ||
AK302625.1 | - | 15 | 1450 | |||
AL833871.1 | - | 6 | 505 | |||
BC144672.1 | - | 14 | 1630 | |||
BC150302.1 | - | 17 | 1999 | 8860399 | ||
DQ309038.1 | - | 15 | 1473 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 17 | 2037 | |||
GenBank mRNA | BC066774.1 | - | 17 | 2011 | ||
BC099913.1 | - | 16 | 2852 | 4838463 | ||
BC125193.1 | - | 16 | 1817 | 40122746 | ||
BC125194.1 | - | 16 | 1817 | 40122749 |