Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 8, 9 | 750 | ||
XM_005256769.1 | XP_005256826.1 | 8, 9 | 747 | |||
XM_005256770.1 | XP_005256827.1 | 7, 8 | 651 | |||
XM_011523993.1 | XP_011522295.1 | 7, 8 | 654 | |||
XM_011523994.1 | XP_011522296.1 | 6, 7 | 618 | |||
XM_011523995.1 | XP_011522297.1 | 8, 9 | 750 | |||
XM_011523996.1 | XP_011522298.1 | 7, 8 | 654 | |||
XM_011523997.1 | XP_011522299.1 | 6, 7 | 388 | |||
GenBank mRNA | AK315818.1 | - | 8, 9 | 737 | ||
BC073889.1 | - | 8, 9 | 719 | 5222492 | ||
BM769737.1 | - | 4, 5 | 174 |
Gene Symbol: LGALS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002308.3 | NP_002299.2 | 7, 8 | 699 | ||
NM_009587.2 | NP_033665.1 | 8, 9 | 795 | |||
NR_024043.1 | 7, 8 | 628 | ||||
XM_006721892.2 | XP_006721955.1 | 6, 7 | 626 | |||
XM_006721893.2 | XP_006721956.1 | 8, 9 | 787 | |||
XM_006721894.3 | XP_006721957.1 | 7, 8 | 662 | |||
XM_006721895.3 | XP_006721958.1 | 6, 7 | 626 | |||
XM_017024623.1 | XP_016880112.1 | 7, 8 | 751 | |||
GenBank mRNA | AB003517.1 | - | 7, 8 | 606 | ||
AB005894.1 | - | 7, 8 | 640 | |||
AB006782.1 | - | 8, 9 | 758 | |||
AB008492.1 | - | 8, 9 | 702 | |||
AK126017.1 | - | 7, 8 | 2049 | |||
AK223232.1 | - | 7, 8 | 663 | |||
AK296031.1 | - | 7, 8 | 570 | |||
AK301624.1 | - | 8, 9 | 665 | |||
BC105942.1 | - | 7, 8 | 623 | 40031795 | ||
BC105944.1 | - | 7, 8 | 623 | 40031802 | ||
BC110340.1 | - | 7, 8 | 640 | 5760999 | ||
Z49107.1 | - | 7, 8 | 668 |
Gene Symbol: LGALS9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042685.1 | NP_001036150.1 | 8, 9 | 742 | ||
GenBank mRNA | AK290263.1 | - | 8, 9 | 746 | ||
AK296659.1 | - | 7, 8 | 659 | |||
BC105943.1 | - | 8, 9 | 722 | 40031793 | ||
BM769345.1 | - | 4, 5 | 174 |