Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 50 | 7154 | ||
GenBank mRNA | AL122111.1 | - | 7 | 655 | ||
BC034223.1 | - | 12 | 1622 | 4838774 |