Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242413.2 | NP_001229342.1 | 17 | 2555 | ||
NM_001282644.1 | NP_001269573.1 | 18 | 2742 | |||
NM_001282645.1 | NP_001269574.1 | 17 | 2683 | |||
NM_001323265.1 | NP_001310194.1 | 18 | 2799 | |||
NM_001323266.1 | NP_001310195.1 | 17 | 2681 | |||
NM_006257.4 | NP_006248.1 | 18 | 2744 | |||
XM_005252496.4 | XP_005252553.1 | 18 | 2844 | |||
XM_006717465.3 | XP_006717528.1 | 20 | 5239 | |||
XM_017016410.1 | XP_016871899.1 | 19 | 5150 | |||
XM_017016411.1 | XP_016871900.1 | 19 | 5178 | |||
GenBank mRNA | BX647657.1 | - | 17 | 2829 | ||
L01087.1 | - | 18 | 2736 | |||
L07032.1 | - | 17 | 2661 |