Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321770.1 | NP_001308699.1 | 2 | 621 | ||
NM_006588.3 | NP_006579.2 | 2 | 621 | |||
NR_135776.1 | 2 | 671 | ||||
XM_017003807.1 | XP_016859296.1 | 2 | 163 | |||
GenBank mRNA | AK297851.1 | - | 2 | 399 | ||
AK298755.1 | - | 2 | 562 | |||
BC058861.1 | - | 2 | 621 | |||
BC125043.1 | - | 2 | 347 | 40115974 | ||
HY118496.1 | - | 1 | 38 |