Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002016.2 | NP_001002016.2 | 12 | 3033 | ||
GenBank mRNA | X66870.1 | - | 12 | 2992 | ||
X76297.1 | - | 12 | 2758 |