Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005246.1 | NP_001005246.1 | 14 | 1879 | ||
XM_008773252.1 | XP_008771474.1 | 17 | 2244 | |||
XM_008773253.1 | XP_008771475.1 | 18 | 2276 | |||
GenBank mRNA | CB750007.1 | - | 1 | 280 |