Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289429.1 | NP_001276358.1 | 5 | 1995 | ||
NM_001289430.1 | NP_001276359.1 | 4 | 1894 | |||
NM_001289431.1 | NP_001276360.1 | 5 | 2068 | |||
NM_001289432.1 | NP_001276361.1 | 5 | 1942 | |||
NM_173735.3 | NP_776096.2 | 4 | 1841 | |||
GenBank mRNA | AK076398.1 | - | 1 | 915 | ||
AK122546.1 | - | 4 | 1770 | |||
AK154476.1 | - | 4 | 1778 | |||
AK160481.1 | - | 5 | 1995 | |||
AK160643.1 | - | 4 | 1767 | |||
BC026384.1 | - | 4 | 1773 | 4168224 |