Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005863.2 | NP_001005863.1 | 14 | 3918 | ||
NM_001005864.3 | NP_001005864.1 | 12 | 1524 | |||
NM_001005865.3 | NP_001005865.2 | 10 | 1537 | |||
NM_001286413.1 | NP_001273342.1 | 14 | 3800 | |||
GenBank mRNA | AF173380.1 | - | 10 | 1376 | ||
AF493235.1 | - | 10 | 1441 | |||
AK030510.1 | - | 10 | 1501 | |||
AK031693.1 | - | 10 | 1471 | |||
AK035576.1 | - | 8 | 905 | |||
AK129324.1 | - | 8 | 4014 | |||
AY246699.1 | - | 10 | 1199 | |||
AY626781.1 | - | 10 | 1534 | |||
AY626782.1 | - | 12 | 1522 | |||
AY626783.1 | - | 13 | 3740 | |||
BC030860.1 | - | 2 | 191 | |||
BC041777.1 | - | 13 | 2583 | 4240274 | ||
BC042206.1 | - | 13 | 2583 | 4240274 | ||
BC043321.1 | - | 12 | 1488 | |||
BC089009.1 | - | 14 | 3800 | 30543240 |