Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181086.3 | NP_851600.1 | 4 | 822 | ||
GenBank mRNA | BC060537.1 | - | 4 | 826 | 6887545 | |
FQ229590.1 | - | 4 | 822 |