Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 9, 10 | 1044 | ||
GenBank mRNA | AF548565.1 | - | 9, 10 | 987 | ||
AK135814.1 | - | 1236 | ||||
BC046963.1 | - | 9, 10 | 982 | |||
BC100666.1 | - | 4, 5 | 350 |